ENA428073876 Enamine

3-methylbenzene-1-sulfonyl isocyanate

  • MDL number MFCD16304425
  • Cas Number : 194543-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. -20

Safety Information

msds https://www.enaminestore.com/catalog/EN300-179418
Availability details for
Quantity
  • Availability
  • Add to Cart