ENA428073887 Enamine

1-(4-fluorophenyl)-5-propyl-1H-pyrazol-4-amine hydrochloride

  • MDL number MFCD28063738
  • Cas Number : 1798685-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-179433

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179433
Availability details for
Quantity
  • Availability
  • Add to Cart