ENA428073888 Enamine

1-(2-aminoethyl)-1H-1,2,3-triazole-4-carboxylic acid hydrochloride

  • MDL number MFCD28023526
  • Cas Number : 1706435-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179434

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179434
Availability details for
Quantity
  • Availability
  • Add to Cart