ENA428073889 Enamine

1,4-difluoro-2-iodo-5-methylbenzene

  • MDL number MFCD11847165
  • Cas Number : 1260826-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179435

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179435
Availability details for
Quantity
  • Availability
  • Add to Cart