ENA428073900 Enamine

2-bromo-5-methyl-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD12404925
  • Cas Number : 1194374-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178389
Availability details for
Quantity
  • Availability
  • Add to Cart