ENA428073902 Enamine

4-(cyanoamino)benzoic acid

  • MDL number MFCD07368594
  • Cas Number : 61700-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179503

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179503
Availability details for
Quantity
  • Availability
  • Add to Cart