ENA428073907 Enamine

2-(2-chloro-3-methylphenyl)acetic acid

  • MDL number MFCD09925311
  • Cas Number : 1000518-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178427

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178427
Availability details for
Quantity
  • Availability
  • Add to Cart