ENA428073910 Enamine

[5-bromo-2-(hydroxymethyl)phenyl]methanol

  • MDL number MFCD15147229
  • Cas Number : 171011-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73-74
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179521

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179521
Availability details for
Quantity
  • Availability
  • Add to Cart