ENA428073918 Enamine

6-amino-5-methylquinolin-2-ol

  • MDL number MFCD20705224
  • Cas Number : 90914-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-179566

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179566
Availability details for
Quantity
  • Availability
  • Add to Cart