ENA428073925 Enamine

1,1'-dimethyl-1H,1'H-[3,4'-bipyrazol]-4-amine dihydrochloride

  • MDL number MFCD28063754
  • Cas Number : 1795437-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179578
Availability details for
Quantity
  • Availability
  • Add to Cart