ENA428073943 Enamine

ethyl 2-[(dimethylamino)methylidene]-3-oxohexanoate

  • Cas Number : 112381-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179679
Availability details for
Quantity
  • Availability
  • Add to Cart