ENA428073947 Enamine

pent-3-en-1-yne

  • Cas Number : 2004-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-179762

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179762
Availability details for
Quantity
  • Availability
  • Add to Cart