ENA428073948 Enamine

4-methylpyridine-3-sulfonyl chloride

  • MDL number MFCD13189159
  • Cas Number : 372198-42-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-179763
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart