ENA428073949 Enamine

3-ethylpentane-3-thiol

  • MDL number MFCD18974479
  • Cas Number : 5827-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-179767

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179767
Availability details for
Quantity
  • Availability
  • Add to Cart