ENA428073950 Enamine

2-chloro-6-fluoro-3-formylbenzonitrile

  • MDL number MFCD24686248
  • Cas Number : 937040-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179768

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179768
Availability details for
Quantity
  • Availability
  • Add to Cart