ENA428073959 Enamine

4,5-dichloro-2-ethylphenol

  • MDL number MFCD24721335
  • Cas Number : 52016-71-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179879

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179879
Availability details for
Quantity
  • Availability
  • Add to Cart