ENA428073960 Enamine

methyl 4-(1H-1,2,3,4-tetrazol-1-yl)benzoate

  • MDL number MFCD02207529
  • Cas Number : 351995-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178649

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178649
Availability details for
Quantity
  • Availability
  • Add to Cart