ENA428073961 Enamine

5-(2-chlorophenyl)-1,2,4-oxadiazol-3-amine

  • MDL number MFCD18866998
  • Cas Number : 1342677-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179883

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179883
Availability details for
Quantity
  • Availability
  • Add to Cart