ENA428073969 Enamine

1-(pyridin-3-yl)-1,2,3,4-tetrahydroisoquinoline

  • MDL number MFCD03825838
  • Cas Number : 28731-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-178657

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178657
Availability details for
Quantity
  • Availability
  • Add to Cart