ENA428073972 Enamine

3-cyclohexylpropanal

  • MDL number MFCD00012280
  • Cas Number : 4361-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179906

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179906
Availability details for
Quantity
  • Availability
  • Add to Cart