ENA428073981 Enamine

2-hydroxy-2-methyl-3-{4-oxo-4H,5H-thieno[3,2-c]pyridin-5-yl}propanoic acid

  • MDL number MFCD25326252
  • Cas Number : 1708229-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179962

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179962
Availability details for
Quantity
  • Availability
  • Add to Cart