ENA428073982 Enamine

2-benzoyl-4-(propan-2-yl)-1,3-thiazole

  • MDL number MFCD18335121
  • Cas Number : 1340453-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178437

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178437
Availability details for
Quantity
  • Availability
  • Add to Cart