ENA428073996 Enamine

1-(2-oxopropyl)pyrrolidine-2,5-dione

  • MDL number MFCD00466564
  • Cas Number : 30057-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178666

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178666
Availability details for
Quantity
  • Availability
  • Add to Cart