ENA428073998 Enamine

1-(3-methylbutyl)-5-(propan-2-yl)-1H-pyrazol-4-amine

  • MDL number MFCD21773966
  • Cas Number : 1522569-34-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-178669
Availability details for
Quantity
  • Availability
  • Add to Cart