ENA428074001 Enamine

1-[(4-ethylphenyl)methyl]-5-methyl-1H-1,2,3-triazol-4-amine

  • MDL number MFCD21947973
  • Cas Number : 1501481-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178670

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178670
Availability details for
Quantity
  • Availability
  • Add to Cart