ENA428074008 Enamine

ethyl 5-oxo-1,2,3,3a,4,5-hexahydropentalene-3a-carboxylate

  • MDL number MFCD23703421
  • Cas Number : 65898-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-180078

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180078
Availability details for
Quantity
  • Availability
  • Add to Cart