ENA428074015 Enamine

(2S)-2-amino-3-ethoxypropanoic acid hydrochloride

  • MDL number MFCD06797557
  • Cas Number : 1311380-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180124
Availability details for
Quantity
  • Availability
  • Add to Cart