ENA428074019 Enamine

4-methoxyoxane-4-carbaldehyde

  • MDL number MFCD24675899
  • Cas Number : 175982-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180131

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180131
Availability details for
Quantity
  • Availability
  • Add to Cart