ENA428074047 Enamine

1,2-dimethoxy-4-(prop-1-en-2-yl)benzene

  • MDL number MFCD20384597
  • Cas Number : 30405-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-180310

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180310
Availability details for
Quantity
  • Availability
  • Add to Cart