ENA428074057 Enamine

2-(1-methyl-1H-pyrazole-4-carbonyl)phenol

  • MDL number MFCD18832625
  • Cas Number : 771481-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180391

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180391
Availability details for
Quantity
  • Availability
  • Add to Cart