ENA428074058 Enamine

4-acetyl-3-(cyclopropylmethoxy)benzonitrile

  • MDL number MFCD26810923
  • Cas Number : 1566804-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180394

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180394
Availability details for
Quantity
  • Availability
  • Add to Cart