ENA428074066 Enamine

1-bromo-2-chloro-4-(trifluoromethoxy)benzene

  • MDL number MFCD11111892
  • Cas Number : 892845-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180442

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180442
Availability details for
Quantity
  • Availability
  • Add to Cart