ENA428074069 Enamine

ethyl 3-(3-bromo-4-chlorophenyl)-3-oxopropanoate

  • MDL number MFCD22200831
  • Cas Number : 112704-82-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178682

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178682
Availability details for
Quantity
  • Availability
  • Add to Cart