ENA428074076 Enamine

N-(pyrimidin-2-yl)prop-2-enamide

  • MDL number MFCD12091200
  • Cas Number : 27179-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178685

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178685
Availability details for
Quantity
  • Availability
  • Add to Cart