ENA428074077 Enamine

5-(benzyloxy)pyrazine-2-carboxylic acid

  • MDL number MFCD20153568
  • Cas Number : 1410093-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180504

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180504
Availability details for
Quantity
  • Availability
  • Add to Cart