ENA428074078 Enamine

4-(2-nitrophenyl)-2-oxobut-3-enoic acid

  • Cas Number : 72323-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180505

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180505
Availability details for
Quantity
  • Availability
  • Add to Cart