ENA428074079 Enamine

1-(4,4,4-trifluorobutyl)-2,5-dihydro-1H-pyrrole-2,5-dione

  • MDL number MFCD24224702
  • Cas Number : 1553221-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-180508
Availability details for
Quantity
  • Availability
  • Add to Cart