ENA428074086 Enamine

2-chloro-6-ethyl-1H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD20526468
  • Cas Number : 1215221-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178686

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178686
Availability details for
Quantity
  • Availability
  • Add to Cart