ENA428074089 Enamine

5-methyl-1lambda4,4-thiazepan-1-one hydrochloride

  • MDL number MFCD28063972
  • Cas Number : 1795189-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180573

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180573
Availability details for
Quantity
  • Availability
  • Add to Cart