ENA428074093 Enamine

1-(2-methylphenyl)-5-propyl-1H-pyrazol-4-amine hydrochloride

  • MDL number MFCD28063974
  • Cas Number : 1798721-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-180578
Availability details for
Quantity
  • Availability
  • Add to Cart