ENA428074099 Enamine

2,4-dihydroxy-N-(2-hydroxyethyl)-3,3-dimethylbutanamide

  • MDL number MFCD24552284
  • Cas Number : 4417-88-3
  • Molecular Weight : 191.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-180594

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180594
Availability details for
Quantity
  • Availability
  • Add to Cart