ENA428074103 Enamine

tert-butyl N-[(2-hydroxyphenyl)methyl]-N-methylcarbamate

  • MDL number MFCD21170282
  • Cas Number : 1487444-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-180600

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180600
Availability details for
Quantity
  • Availability
  • Add to Cart