ENA428074110 Enamine

4-[3-(trifluoromethyl)phenyl]cyclohexan-1-one

  • MDL number MFCD25323932
  • Cas Number : 40503-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180624

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180624
Availability details for
Quantity
  • Availability
  • Add to Cart