ENA428074112 Enamine

5-methyl-N-(propan-2-yl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD00124189
  • Cas Number : 62347-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180634

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180634
Availability details for
Quantity
  • Availability
  • Add to Cart