ENA428074116 Enamine

4-(2-methyl-1,3-dioxolan-2-yl)-1,2-dihydropyridin-2-one

  • MDL number MFCD24597925
  • Cas Number : 192819-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180648

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180648
Availability details for
Quantity
  • Availability
  • Add to Cart