ENA428074123 Enamine

5-cyano-2-hydroxybenzoic acid

  • MDL number MFCD16999178
  • Cas Number : 10435-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210-212
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-180726

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180726
Availability details for
Quantity
  • Availability
  • Add to Cart