ENA428074125 Enamine

methyl 2-amino-5-fluoro-3-nitrobenzoate

  • MDL number MFCD12159895
  • Cas Number : 328547-11-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-180736

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180736
Availability details for
Quantity
  • Availability
  • Add to Cart