ENA428074130 Enamine

1-[(4-fluorophenyl)methyl]-5-methyl-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD06797392
  • Cas Number : 885950-27-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180745

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180745
Availability details for
Quantity
  • Availability
  • Add to Cart