ENA428074131 Enamine

3-(2-aminoethyl)cyclobutan-1-ol hydrochloride

  • MDL number MFCD28063986
  • Cas Number : 1789007-32-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-180748

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180748
Availability details for
Quantity
  • Availability
  • Add to Cart