ENA428074143 Enamine

2-(1H-pyrazol-3-yl)-1,3-thiazole

  • MDL number MFCD00105072
  • Cas Number : 166196-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-154
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-180806

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180806
Availability details for
Quantity
  • Availability
  • Add to Cart